Learn More
Liebherr™ NoFrost Comfort Freezer
Say goodbye to defrosting. Liebherr™ NoFrost Comfort Freezer eliminates ice formation. Rapidly freeze newly introduced items and efficiently maintain a stable frozen environment. Features clear drawers with glass shelves. Available in multiple sizes to accommodate space requirements and storage needs.
Marca:  Liebherr™ GNP 2713
Detalles adicionales : Peso : 0.00010kg
Código de producto. 15123449
  • 1375.00€ / cada uno
Fecha estimada de envío: 12-08-2024
para ver el stock
Añadir a la cesta
Descripción
Descripción
Rapid, efficient freezing with no icing
  • NoFrost
    • Products are efficient frozen with chilled recirculating air
    • Any humidity is expelled so the freezer is always free of ice and nothing frosts over
    • Eliminates the need for defrosting
  • SuperFrost
    • Automatic feature engages when new products are loaded
    • Quickly reduces the temperature to -32 °C to quickly and efficiently freeze products
    • Switches back to normal mode when frozen (max 65 hours) to save electricity
  • FrostSafe
    • Drawers are closed all around so the cold does not quickly dissipate quickly when appliance is opened
    • Transparent drawers allows content identification without opening the drawer
Customizable interior and exterior
  • Removable drawers allow you to load items directly onto the glass shelves beneath them
  • VarioSpacesystem: removable drawers and shelves let you accommodate larger items
  • Reversible door hinges permit the door to open from either side
  • Adjustable feet for use on uneven floors and included rollers for easy transport
Easy and convenient to use
  • Electronic temperature control with MagicEye digital touchpad display
  • Audible alarm when the door has been left open or the temperature rises
  • Slimline door handle with integrated opening mechanism for easy, nearly automatic door opening
  • Integrated door lock to keep contents safe and secure
  • Premier A++ energy efficiency rating and eco-friendly refrigerant