Learn More
NOL6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530807
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: IMR32, U87-MG, mouse brain. Predicted reactivity: Mouse (89%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The nucleolus is a dense subnuclear membraneless organelle that assembles around clusters of rRNA genes and functions in ribosome biogenesis. This gene encodes a nucleolar RNA-associated protein that is highly conserved between species. RNase treatment of permeabilized cells indicates that the nucleolar localization is RNA dependent. Further studies suggest that the protein is associated with ribosome biogenesis through an interaction with pre-rRNA primary transcripts. Alternative splicing has been observed at this locus and two splice variants encoding distinct isoforms have been identified.Especificaciones
NOL6 | |
Polyclonal | |
Unconjugated | |
NOL6 | |
AA410091; bA311H10.1; LOW QUALITY PROTEIN: nucleolar protein 6; Nol6; Nrap; nucleolar protein 6; nucleolar protein 6 (RNA-associated); nucleolar protein family 6; nucleolar protein family 6 (RNA-associated); nucleolar RNA-associated protein; UTP22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230082, 65083 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8R5K4, Q9H6R4 | |
NOL6 | |
Recombinant fragment corresponding to a region within amino acids 933 and 1146 of Human NOL6 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |