Learn More
Novus Biologicals™ Non-human Primate Apolipoprotein A-I/ApoA1 ELISA Kit
ELISA Kit
Marca: Novus Biologicals™ NBP3-11735
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
0.63 to 40 ng/mL | |
ELISA | |
ELISA Kit | |
Non-human Primate | |
335 | |
1 Kit (1 Plate) | |
Storage of components varies. See protocol for specific instructions. | |
Non-human Primate |
0.2 ng/mL | |
ELISA (Standard) | |
Serum, Plasma, Cell Culture Supernatant | |
ELISA | |
APOA1 | |
Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, MGC117399 |