Learn More
Nope Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA552580
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: VEDRAEVHSL MGGGVSEGRS HSKRKISWAQ PSGLSWAGSW AGCELPQAGP RPALTRALLP PAGTGQTLLL QALVYDAIKG NGRKKSPPAC RNQVEAEVIV HSDFSASNGN PDLHLQDLEP EDPLPPEAPD LISGVGD Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
Nope Polyclonal antibody specifically detects Nope in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Especificaciones
Nope | |
Polyclonal | |
Unconjugated | |
IGDCC4 | |
amplimer WI-16786, mouse homolog; amplimer WI-18508, mouse homolog; DDM36; hDDM36; Igdcc4; Immunoglobulin superfamily DCC subclass member 4; immunoglobulin superfamily, DCC subclass, member 4; Kiaa1628; likely ortholog of mouse neighbor of Punc E11; LOC57722; neighbor of Punc E11; neighbor of Punc e11 protein; Nope; NOPEFLJ42051; Protein DDM36 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57722 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDY8 | |
IGDCC4 | |
Recombinant protein corresponding to Human Nope | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only