Learn More
NOR-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA124536
Detalles adicionales : Peso : 0.01000kg
Descripción
PA1-24536 detects NOR-1 in Human samples. PA1-24536 has been successfully used in Immunohistochemistry (paraffin) procedures. PA1-24536 immunogen corresponds to Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 65/75%. Rat: 65/75%.
NOR-1, a NR4 nerve growth factor-like receptor, has been shown to affect neuronal development, T-cell apoptosis, and cancer development. NOR-1 binds to NBRE (NGFI-B response element) and regulates the pro-opiomelanocortin (POMC) gene. NOR-1 is induced rapidly but transiently by a variety of stimuli. Downregulation of NOR-1 and other NGFI-B family members is correlated with drug addiction-prone behavior of Lewis rats. Three human protein isoforms have been documented: NOR-1, NOR-1beta, and NOR-1 3′ variant.Especificaciones
NOR-1 | |
Polyclonal | |
Unconjugated | |
Q92570 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8013 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
NR4A3 | |
NR4A3 | |
Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 65/75%. Rat: 65/75%. | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |