missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTCH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA523181
Detalles adicionales : Peso : 0.01000kg
Descripción
The notch gene belongs to a family of epidermal growth factor (EGF) like homeotic genes, which encode transmembrane proteins with a variable number of cysteine-rich EGF-like repeats in the extracellular region. Four notch genes have been described in mammals: Notch1, Notch2, Notch3 and Notch4(Int-3), which have been implicated in the differentiation of the nervous system and other structures. The EGF-like proteins Delta and Serrate have been identified as ligands of Notch1. Mature Notch proteins are heterodimeric receptors derived from the cleavage of Notch pre-proteins into an extracellular subunit (NEC) containing multiple EGF-like repeats and a transmembrane subunit including intracellular region (Ntm).Especificaciones
NOTCH1 | |
Polyclonal | |
Unconjugated | |
Notch1 | |
9930111A19Rik; AOS5; AOVD1; Drosophila Notch homolog 1 (controlling the the ectodermal and neural cell fate in Drosophila); hN1; lin-12; major type A protein; Mis6; Motch; Motch A; mT14; N1; neurogenic locus notch homolog protein 1; NEXT; NICD; NOTCH; notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch gene homolog 1; Notch homolog 1, translocation-associated; notch receptor 1; Notch1; p300; RP23-306D20.12; RP23-306D20.12-001; Tan1; Translocation-associated notch protein TAN-1; transmembrane receptor Notch1 | |
Rabbit | |
Protein G | |
RUO | |
18128, 25496, 480676, 4851, 767866 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
P46531, Q01705, Q07008 | |
Notch1 | |
Within the range of amino acids 1755-1780 of human Notch1 | |
100 μg | |
Primary | |
Bovine, Canine, Human, Mouse, Rat | |
Antibody | |
IgG |