Learn More
Novobiocin sodium salt, Thermo Scientific™ Código de producto.: 15413619
Novobiocin sodium salt, Thermo Scientific™
Código de producto. 15413619
Código de producto. 15413619
Marca: Thermo Scientific Alfa Aesar J60928.09
Solicitar formato a granel o personalizado
Novobiocin sodium salt, CAS # 1476-53-5, is a topoisomerase II inhibitor capable of inhibiting DNA synthesis. This compound is produced by Streptomyces nieveus and shows antibiotic activity.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Novobiocin sodium salt, first isolated from Streptomyces nieveus, is a topoisomerase II inhibitor. It is a coumarin compound showing antibiotic activity
• Novobiocin sodium salt can inhibit DNA synthesis by inhibiting the topoisomerase II enzyme. Its antibacterial activity arises from competitive inhibition of the ATPase reaction catalyzed by the GyrB subunit
Application
• Novobiocin sodium salt is used to inhibit the activity of retroviral RNA-dependent DNA polymerase in laboratory experiments
• It is also used as an inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase.
• Novobiocin sodium salt shows anti-staphylococcal activity against methicillin-resistant Staphylococcus aureus, a gram-positive bacterium
• It permits the relaxation of negative DNA supercoils and has been used to generate positively supercoiled DNA
• This compound inhibits the chaperone activity of Hsp90
TRUSTED_SUSTAINABILITY
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
5 Resultados encontrados
Número de lote Tipo de certificado Fecha Catalog Number
Número de loteZ24K043 Tipo de certificadoCertificado de análisis Fecha21/01/2025 Catalog NumberJ60928.03,J60928.06,J60928.09
Número de loteW19K020 Tipo de certificadoCertificado de análisis Fecha27/09/2024 Catalog NumberJ60928.03,J60928.06,J60928.09
Número de loteQ23K037 Tipo de certificadoCertificado de análisis Fecha11/06/2024 Catalog NumberJ60928.03,J60928.06,J60928.09
Número de loteQ05K033 Tipo de certificadoCertificado de análisis Fecha18/04/2024 Catalog NumberJ60928.03,J60928.06,J60928.09
Número de loteP02K007 Tipo de certificadoCertificado de análisis Fecha25/03/2024 Catalog NumberJ60928.03,J60928.06,J60928.09
RUO – Research Use Only
Título del producto
Novobiocin sodium salt, Thermo Scientific™ >