Learn More
NOX4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553304
Additional Details : Peso : 0.02000kg
Description
Immunogen sequence: ISLNRTSSQN ISLPEYFSEH FHEPFPEGFS KPAEFTQHKF VKICMEEPRF QANFPQTWLW ISGPLCLYCA ERLYRYIRSN KPVTIISVIS HPSDVMEIRM VKENFKARPG QYI Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.
NOX4 Polyclonal antibody specifically detects NOX4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
NOX4 | |
Polyclonal | |
Unconjugated | |
Nox4 | |
AI648021; kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX; KOX1; KOX-1; NADPH oxidase 4; NOX 4; NOX4; NOX-4; renal NAD(P)H-oxidase; RENOX; superoxide-generating NADPH oxidase 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50507 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NPH5 | |
Nox4 | |
Recombinant protein corresponding to Human NOX4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only