Learn More
Noxa Antibody (S07-3H6), Novus Biologicals™
Rabbit Monoclonal Antibody
Marca:  Novus Biologicals NBP3-19820-50ul
 Ver más versiones de este producto
Código de producto. 18605763
  • 421.00€ / 50 microlitros
Descripción
Noxa Monoclonal antibody specifically detects Noxa in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Noxa
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
S07-3H6
Unconjugated
adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
A synthetic peptide of human Noxa (Uniprot # Q13794)
50 μg
Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Mitochondrial Mediated Pathway, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors
5366
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones