missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOXA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA526572
Detalles adicionales : Peso : 0.01000kg
Descripción
NADPH oxidases (NOXs) catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species (ROS). The NOX activator NOXA1 can stimulate both NOX1 and NOX2, but it appears to be more effective in activating NOX1.Especificaciones
NOXA1 | |
Polyclonal | |
Unconjugated | |
NOXA1 | |
antigen NY-CO-31; inhibitory NADPH oxidase activator 1; NADPH oxidase activator 1; NCF2-like protein; Nox activator 1; Noxa1; NY-CO-31; p51NOX; p51-nox; P67phox-like factor; predicted NADPH oxidase activator 1; SDCCAG31; serologically defined colon cancer antigen 31 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10811 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q86UR1 | |
NOXA1 | |
KLH conjugated synthetic peptide between 449-478 amino acids from the C-terminal region of human NOXA1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |