missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPSR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA533986
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster (100%); Elephant, Panda, Bovine, Bat, Pig, Turkey, Chicken (94%); Dog, Opossum, Lizard (89%); Horse, Platypus (83%). Proteinase K treatment is required for antigen retrieval.
May be active in signaling pathway in an autocrine or paracrine fashion in several tissues. Receptor for neuropeptide S, it may mediate its action, such as inhibitory effects, on cell growth. Involved in pathogenesis of asthma and other IgE-mediated diseases.Especificaciones
NPSR1 | |
Polyclonal | |
Unconjugated | |
NPSR1 | |
9330128H10Rik; ASRT2; G protein-coupled receptor 154; G protein-coupled receptor PGR14; Gpr154; GPRA; G-protein coupled receptor 154; G-protein coupled receptor for asthma susceptibility; G-protein coupled receptor PGR14; MVTR; Neuropeptide S receptor; neuropeptide S receptor 1; NPSR; Npsr1; Pgr14; RGD1564154; vasopressin receptor-related receptor 1; vasotocin receptor; VRR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
300458, 319239, 387129 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P0C0L6, Q6W5P4, Q8BZP8 | |
NPSR1 | |
Synthetic 18 amino acid peptide from 2nd extracellular domain of human NPSR1. | |
50 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |