missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NTRK2 (TRKB), His Tag Recombinant Protein

Código de producto. p-7181589
Click to view available options
Cantidad:
10 μg
Tamaño de la unidad:
10 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11503477

Marca: Invitrogen™ PV3616

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic.

NTRK2 (TRKB) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. NTRK2 is the receptor for brain-derived neurotrophic factor (BDNF). Together, NTRK2 and BDNF regulate both short-term synaptic functions and long-term potentiation of brain synapses. TrkB is involved in the development and maintenance of the nervous system. Moreover, Trk B is a tyrosine kinase gene highly related to Trk A. Trk B expression is confined to tissues within the central and peripheral nervous systems. The brain-derived neurotrophic factor (BDNF) and NT-3, but not NGF, can induce rapid phosphorylation on tyrosine of Trk B gp145, one of the receptors encoded by Trk B, although BDNF elicits a response at least two orders of magnitude greater than NT-3. TrkB is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signaling through TrkB leads to cell differentiation. Mutations in the TrkB gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for the TrkB gene.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q16620
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50mM tris HCl with 300mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol and no preservative; pH 7.5
ID de gen (Entrez) 4915
Peso molecular 40.9 kDa
Nombre Human NTRK2 (TRKB), His Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 10 μg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen AI848316; BDNF/NT-3 growth factors receptor; BDNF-tropomyosine receptor kinase B; C030027L06Rik; EC 2.7.10.1; GP145 TrkB; GP145TrkB; GP145-TrkB; GP145-TrkB/GP95-TrkB; GP95 TrkB; GP95-TrkB; hyperphagia; kinase TrkB; neural receptor protein-tyrosine kinase (trkB); neurotrophic receptor tyrosine kinase 2; neurotrophic tyrosine kinase receptor type 2; neurotrophic tyrosine kinase, receptor, type 2; neurotrophic tyrosine receptor kinase type 2; NTRK 2; Ntrk2; OTTHUMP00000021573; OTTHUMP00000021574; OTTHUMP00000021576; OTTHUMP00000021577; RATTRKB1; Tkrb; Trk B; TRKB; trk-B; trkB protein; trkB tyrosine kinase; TRKB1; Tropomyosin-related kinase B; tyrosine kinase receptor B
Nombre común TrkB
Símbolo de gen Ntrk2
Familia Quinasa Neurotrophic Receptor (NTRK) Kinase Family
Longitud de la proteína 526-838
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-tag
Secuencia 526-838
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Entrada del ensayo Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity
Grupo de cinasas Tyrosine Kinases
Condiciones de envío Dry Ice
Compatibilidad de sistemas LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Entrada diana NTRK2
Gen diana neurotrophic tyrosine kinase, receptor, type 2
Sistema de expresión Insect cells
Familia de proteínas Kinases & Inhibitors
Forma de proteína Recombinant, Cytoplasmic
Formulario Liquid
Subtipo de proteína Receptor Tyrosine Kinases
Categoría de investigación Signal Transduction
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado