missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NXT1 293T Cell Overexpression Lysate (Denatured), Abnova
Marca: Abnova H00029107-T01.100uL
Detalles adicionales : Peso : 0.00010kg
Descripción
Western Blotting
Especificaciones
Western Blot | |
NXT1 293T Cell Transient Overexpression Lysate(Denatured) | |
15.51 | |
pCMV-NXT1 full-length | |
100 μL | |
Human |
293T | |
BC000759 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-NXT1 antibody (H00029107-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UL NXT1 293T Cell Transient Overexpression Lysa
Signal Word
- Atención
Hazard Category
- Sensibilización cutánea Categoría 1
Hazard Statement
- H317-Puede provocar una reacción alérgica en la piel.
Precautionary Statement
- P261-Evitar respirar el polvo/el humo/el gas/la niebla/los vapores/el aerosol.
- P272-Las prendas de trabajo contaminadas no podrán sacarse del lugar de trabajo.
- P280-Llevar guantes/prendas/gafas/máscara de protección.
Supplemental information
- MIXTURE LIST-Contém: Mercaptoethanol