missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Occludin Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech CL488-13409-100UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Occludin Polyclonal antibody specifically detects Occludin in Human, Mouse, Rat samples. It is validated for IFEspecificaciones
Occludin | |
Polyclonal | |
CoraLite 488 | |
BC013596 | |
Antigen affinity purification | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunofluorescence | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
Rabbit | |
100 μL | |
4950 | |
Store at -20°C | |
Liquid |