Learn More
Kern Sohn OIML Class E2 Weight Safety Set
Safety set contains one test weight for testing the upper end of the weighing range of your balance and one test weight for testing the lower end.
Marca:  Kern Sohn 315-107-104
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17321953
  • 239.70€ / 1 set
Fecha estimada de envío: 15-01-2025
para ver el stock
Explore más ofertas especiales
Añadir a la cesta
Descripción
Descripción
  • These KERN Safety Sets each contain one test weight for testing the sensitivity, i.e. the correct adjustment of your balance as well as a small test weight to test the lower end of the weighing range, the so-called minimum sample weight
  • As an option, the KERN Safety Set has space for another test weight, for testing your balance at a weight which is relevant for you
  • Useful accessories which have been selected to suit that particular KERN Safety Set, such as, for example, special gloves, tweezers, weight grips, brushes, etc. will assist you in handling your test weights properly
  • Stored in the practical protective case next to your balance, you can check and ensure the high precision of your balance at any time
Especificaciones
Especificaciones
E2
Acero inoxidable
10, 100 g
Caja protectora