Learn More
Kern Sohn OIML Class E2 Weight Safety Set
Safety set contains one test weight for testing the upper end of the weighing range of your balance and one test weight for testing the lower end.
Marca: Kern Sohn 315-102-187
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- These KERN Safety Sets each contain one test weight for testing the sensitivity, i.e. the correct adjustment of your balance as well as a small test weight to test the lower end of the weighing range, the so-called minimum sample weight
- As an option, the KERN Safety Set has space for another test weight, for testing your balance at a weight which is relevant for you
- Useful accessories which have been selected to suit that particular KERN Safety Set, such as, for example, special gloves, tweezers, weight grips, brushes, etc. will assist you in handling your test weights properly
- Stored in the practical protective case next to your balance, you can check and ensure the high precision of your balance at any time
Especificaciones
E2 | |
Acero inoxidable | |
100 mg, 2 g | |
Caja protectora |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.