Learn More
Kern Sohn OIML Class F2 Weight Set, Wooden Box
Weight set constructed of finely turned stainless steel and housed in a wooden box.
Marca:  Kern Sohn 334-04
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17312133
  • 387.60€ / 1 set
Fecha estimada de envío: 13-01-2025
para ver el stock
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Knob shape
  • Finely turned stainless steel
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic box
Specifications
Specifications
F2
Acero inoxidable
Wood
1 a 200 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.