Learn More
Kern Sohn OIML Class F2 Weight Set, Wooden Box
Weight set constructed of finely turned stainless steel and housed in a wooden box.
Marca: Kern Sohn 334-03
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Knob shape
- Finely turned stainless steel
- Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic box
Especificaciones
F2 | |
Acero inoxidable | |
Wood | |
De 1 a 100 g |