missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPRK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA533860
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Dog, Horse, Rabbit (100%); Gorilla, Mouse, Rat, Elephant, Guinea pig (95%); Bat, Bovine, Panda (89%); Opossum (84%). Please note that this antibody produces faint nuclear background staining in some tissues.
This gene encodes an opioid receptor, which is a member of the 7 transmembrane-spanning G protein-coupled receptor family. It functions as a receptor for endogenous ligands, as well as a receptor for various synthetic opioids. Ligand binding results in inhibition of adenylate cyclase activity and neurotransmitter release. This opioid receptor plays a role in the perception of pain and mediating the hypolocomotor, analgesic and aversive actions of synthetic opioids. Variations in this gene have also been associated with alcohol dependence and opiate addiction. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. A recent study provided evidence for translational readthrough in this gene, and expression of an additional C-terminally extended isoform via the use of an alternative in-frame translation termination codon.Especificaciones
OPRK1 | |
Polyclonal | |
Unconjugated | |
OPRK1 | |
kappa opioid receptor; kappa-type opioid receptor; KOPR1; KOR; K-OR-1; KOR-1; MSL-1; Opiate receptor, kappa-1; opioid receptor kappa 1; opioid receptor, kappa 1; opioid receptor, kappa 2; OPRK; OPRK1; Oprk2; Ork1; R21; Ror-d | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100054492, 100354170, 29335, 486951, 4986 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P34975, P41145 | |
OPRK1 | |
Synthetic 19 amino acid peptide from 2nd extracellular domain of human Kappa Opioid Receptor. | |
50 μg | |
Primary | |
Canine, Equine, Human, Rabbit, Rat | |
Antibody | |
IgG |