Learn More
OPRL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA3029
Detalles adicionales : Peso : 0.00900kg
Descripción
IHC (P) analysis shows positive staining of NOP in human enteric ganglion. WB analysis shows NOP in glycoprotein-enriched fractions from mock transfected or NOP overexpressing 293 cells. While NOP has a theoretical MW of ∽ 41kD, the observed band in a WB runs at ∽72kD due to enrichment of the glycosylated receptor.
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. The nociceptin receptor is a G-protein coupled opioid receptor that functions as receptor for the endogenous neuropeptide nociceptin. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors. Signaling via G proteins mediates inhibition of adenylate cyclase activity and calcium channel activity. Arrestins modulate signaling via G proteins and mediate the activation of alternative signaling pathways that lead to the activation of MAP kinases. Plays a role in modulating nociception and the perception of pain. Plays a role in the regulation of locomotor activity by the neuropeptide nociceptin.Especificaciones
OPRL1 | |
Polyclonal | |
whole serum with 0.05% sodium azide | |
P41146 | |
OPRL1 | |
KLH conjugated Cys-VRSIAKDVALACKTSETVPRPA | |
RUO | |
4987 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
OPRL1 | |
NOP, Nociceptin Receptor, Opiate Receptor-Like 1, NOPR, OPPRL1, Orphanin FQ Receptor, Kappa3-Related Opioid Receptor, Kappa-Type 3 Opioid Receptor, KOR-3, OOR, ORL1, LC132 Receptor-Like, NOCIR | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |