missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OV-6 Antibody (OV-6) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB2020JF585
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
OV-6 Monoclonal antibody specifically detects OV-6 in Human,Rat samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOFEspecificaciones
OV-6 | |
Monoclonal | |
Janelia Fluor 585 | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Rat | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
OV-6 | |
50mM Sodium Borate | |
Carcinogen-treated rat nodular hepatocytes | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |