Learn More
Honeywell Safety Products™ Overspec Safety Glasses
Click to view available options
:
Tipo de lente: Resistentes a impactos
Descripción
- Over-the-glass coverage, offering workers full protection while wearing their own prescription eyewear.
- Overspec offers all day coverage for a variety of workers and a wide range of prescription eyewear styles.
- Bendable temples fit virtually all users.
- Extremely comfortably and secure.
- Temples curved outward to minimize temple interference with prescription eyewear .
- Bendable temples to accommodate their face, head size and prescription frames.
- Ratcheting temple (3 positions) to further customize the fit.
Especificaciones
Especificaciones
| Certificaciones/conformidad | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number C899.1B-D, EC Attestation Number C899.1B-D |
| Revestimiento de lente | Anti-fog, Impact-resistant |
| Tipo de producto | Gafas de seguridad |
| Para utilizar con (aplicación) | Agriculture, Automotive, Building, Construction, Catering, Fishing, Food, Green Spaces, Laboratory, Logistics, Maintenance, Medical, Pharmaceutical, Service, Transportation and Wood industries. |
| Color de marco | Azul, negro |
| N.° por paquete | 1 Ea. |
| Lens Material | Policarbonato |
| Material del bastidor | Nailon |