Learn More
Tocris Bioscience™ OXF BD 02
Selective BRD4(1) inhibitor
Marca: Tocris Bioscience™ 4928/10
Detalles adicionales : CAS : 1429129-68-9 Peso : 0.00970kg
Descripción
OXF BD 02 is a selective inhibitor of the first bromodomain of BRD4 (BRD4(1)) (IC50 = 382 nM). Exhibits 2-3-fold selectivity for BRD4(1) over the CBP bromodomain and has little affinity for a range of other bromodomains. Reduces viability of lung adenocarcinoma cell lines and attenuates proliferation of MV-4-11 leukemia cells. Cell permeable.Especificaciones
>98% | |
10 mg |
For Research Use Only