missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y12/P2RY12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Marca: Novus Biologicals NBP2-33870-25ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
P2Y12/P2RY12 Polyclonal specifically detects P2Y12/P2RY12 in Human, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
| P2Y12/P2RY12 | |
| Polyclonal | |
| Western Blot - reported in scientific literature (PMID:31968618)., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence -validated from a verified customer review, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunohistochemistry-Frozen -validated from a verified customer review | |
| Q9H244 | |
| P2RY12 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KSFRNSLISMLKCPNSATSLSQDNRKKEQDGGDPNEETPM | |
| 25 μL | |
| GPCR | |
| 64805 | |
| Human, Feline | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human P2Y12/P2RY12 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido