missing translation for 'onlineSavingsMsg'
Learn More

p38 MAPK gamma Monoclonal Antibody (10E1), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA1100

Código de producto. 13287628

  • 512.00€ / 100 microgramos

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

MA1-100 has been successfully used in Western blot and immunohistochemistry applications and reacts with human and mouse samples. MA1-100 specifically recognizes p38-gamma, and does not react with alpha, beta, or delta isoforms.

MAPK12 (p38 gamma) is a serine/threonine kinase involved in many different cellular processes. It is activated during hypoxia and gamma-irradiation-induced G(2) arrest. Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. MAPK12 functions as a signal transducer during differentiation of myoblasts to myotubes.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

p38 MAPK gamma
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
O08911, P53778
Mapk12
Protein expressed in 293T cell transfected with human MAPK12 expression vector
100 μg
Primary
Canine, Human, Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
10E1
Unconjugated
Mapk12
AW123708; ERK3; ERK5; erk6; ERK-6; etID309866.18; Extracellular signal-regulated kinase 6; fa05c12; LOW QUALITY PROTEIN: mitogen-activated protein kinase 12; mitogen-activated protein kinase 12; MAP kinase 12; MAP kinase p38 gamma; MAPK 12; mapk12; mapk12a; mitogen activated protein kinase 12; mitogen-activated protein kinase 12; mitogen-activated protein kinase 12a; mitogen-activated protein kinase 3; mitogen-activated protein kinase p38 gamma; OTTHUMP00000196671; P38gamma; p38-gamma; Prkm12; SAP kinase-3; sapk3; SAPK-3; stress activated protein kinase 3; stress-activated protein kinase 3; stress-activated protein kinase-3; wu:fa05c12; zgc:101695
Mouse
Protein A
RUO
29857, 607023, 6300
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales