Learn More
Tocris Bioscience™ P3S
Reagent for modifying RNA at 2-OH position, for SHAPE-MAP experiments
Marca:  Tocris Bioscience™ 7974/50
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18055132
  • 326.47€ / 50 miligramos
Fecha estimada de envío: 12-08-2024
para ver el stock
Descripción
Descripción
P3S is a pyridine sulfonyl triazole able to modify RNA via RNA 2-OH sulfonylation. P3S can be used for RNA SHAPE-MaP experiments in cell lines or live bacteria. It is stable in aqueous media (with half-lives of several hours) even at room temperature.
Especificaciones
Especificaciones
3-((1H-1,2,4-Triazol-1-yl)sulfonyl)pyridine
DNA, RNA and Protein Synthesis Compounds
95%
50 mg
C7H6N4O2S