Learn More
Tocris Bioscience™ P3S
Reagent for modifying RNA at 2-OH position, for SHAPE-MAP experiments
Marca: Tocris Bioscience™ 7974/50
Detalles adicionales : Peso : 0.00970kg
Descripción
P3S is a pyridine sulfonyl triazole able to modify RNA via RNA 2-OH sulfonylation. P3S can be used for RNA SHAPE-MaP experiments in cell lines or live bacteria. It is stable in aqueous media (with half-lives of several hours) even at room temperature.Especificaciones
3-((1H-1,2,4-Triazol-1-yl)sulfonyl)pyridine | |
DNA, RNA and Protein Synthesis Compounds | |
95% |
50 mg | |
C7H6N4O2S |