missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p57 Kip2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-28707
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
p57 Kip2 Polyclonal antibody specifically detects p57 Kip2 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Especificaciones
| p57 Kip2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) | |
| Beckwith-Wiedemann syndrome, BWS, cyclin-dependent kinase inhibitor 1C, cyclin-dependent kinase inhibitor 1C (p57, Kip2), Cyclin-dependent kinase inhibitor p57, KIP2BWCR, p57, p57Kip2, WBS | |
| The immunogen recognized by this antibody maps to a region between residue 266 and 316 of human cyclin-dependent kinase inhibitor p57 using the numbering given in entry NP_000067.1 (GeneID 1028). | |
| 100 μg | |
| Cell Cycle and Replication | |
| 1028 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot (Negative) | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-5 μg/mg lysate, Western Blot (Negative) | |
| NP_000067.1 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction