missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAK1 Recombinant Human Protein
PAK1 is a serine/threonine kinase.
Marca: Invitrogen™ PV4030
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PAK1 is a target for restenosis, cancer, and immunosuppression treatment.
Especificaciones
NP_002567 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
PAK1, PAK alpha | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
88.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |