missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pannexin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen 422900
341.60 EUR hasta el 2025-03-16
Use el código promocional "24070" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Gap junctions are channel-forming structures that allow direct metabolic and electrical communication between adjacent cells of almost all types in mammalian tissues. In the human body, they are absent only in adult skeletal muscle cells and some circulating blood cells. A gap junction is formed two hemichannels, one in each of the neighboring cells, composed of six subunits. In mice and humans, at least 20 connexin and 3 pannexin genes encode gap junction proteins. Connexins are only found in chordates, while pannexins are present in both chordate and invertebrate genomes. Pannexins, previously known as innexins, are predicted to have four transmembrane regions, two extracellular loops, one intracellular loop, and intracellular N- and C-termini. Both human and mouse genomes contain three pannexin-encoded genes. Pannexin 2 (Px2, PANX2) appears to be a brain specific gene, and is abundantly expressed in the central nervous system, as is pannexin 1. In many neuronal cell populations, including hippocampus, olfactory bulb, cortex, and cerebellum, pannexin 1 and pannexin 2 are co-expressed; in other brain regions such as white matter, only pannexin 1-positive cells are found.
Especificaciones
Pannexin 2 | |
Polyclonal | |
Unconjugated | |
PANX2 | |
h PX2; hPANX2; pannexin 2; pannexin-2; PANX2; Px2; si:ch211-192n14.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
362979, 406218, 56666 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P60571, Q6IMP4, Q96RD6 | |
PANX2 | |
Synthetic peptide derived from an internal region of human, mouse, and rat pannexin 2, which is identical to predicted bovine and dog sequence | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-5 of 11
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Pannexin 2 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido