Learn More
Motic™ Panthera Cloud Binocular Digital Microscope
The Panthera Cloud has smartly been created to make your workflow easier with strong Ultra Contrast optics.
Marca: Motic™ 1100104601181
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- 25° inclined binocular Siedentopf head
- Rotates 360°
- SmartCam 5 MP camera
- Strong optical performance with Ultra Contrast (UC) optics
- Diopter Adjustment: ±4
Especificaciones
WF 10X/22 mm | |
Kohler Illumination | |
Plan Achromat Objectives UC (4X, 10X, 40X(S), 100X(S-oil)) |
Binocular | |
10X |