Learn More
DiscoverX PathHunter™ eXpress ACVR1C/ACVR2B Dimerization Assay
Click to view available options
Cantidad:
200 dp (2 x 96-pocillos)
Tamaño de la unidad:
1 set
Descripción
The PathHunter™ ACVR1C/ACVR2B Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. activin B) induced dimerization of ACVR1C/ACVR2B receptors. This product uses EFC technology. The cell line included overexpresses ACVR2B target protein fused to PK alongside ACVR1C target protein fused to EA. Upon ligand-induced dimerization of ACVR1C/ACVR2B , detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | ACVR1C/ACVR2B Dimerization |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Agonista primario | Activin B |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | ACVR1C/ACVR2B |
| Línea de células | U2OS |
| Objetivo | ACVR1C/ACVR2B |