Learn More
DiscoverX PathHunter™ eXpress BMPR1A/ACVR2 Dimerization Assay
Click to view available options
Cantidad:
1000 dp (10 x 96-pocillos)
Tamaño de la unidad:
1 set
Descripción
The PathHunter™ BMPR1A/ACVR2 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. BMP-4) induced dimerization of BMPR1A/ACVR2 receptors. This product uses EFC technology. The cell line included overexpresses BMPR1A target protein fused to PK alongside ACVR2 target protein fused to EA. Upon ligand-induced dimerization of BMPR1A/ACVR2 , detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | BMPR1A/ACVR2 Dimerization |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Agonista primario | BMP-4 |
| Cantidad | 1000 dp (10 x 96-pocillos) |
| Entrada diana | BMPR1A/ACVR2 |
| Línea de células | U2OS |
| Objetivo | BMPR1A/ACVR2 |