Learn More
DiscoverX PathHunter™ eXpress CMKLR1 Activated GPCR Internalization Assay
Click to view available options
Cantidad:
200 dp (2 x 96-pocillos)
Tamaño de la unidad:
1 set
Descripción
The PathHunter™ CMKLR1 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. chemerin) based internalization of CMKLR1 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged CMKLR1. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting CMKLR1-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | GPCR |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | CMKLR1 |
| Objetivo | CMKLR1 |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.