missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress CXCR2 Activated GPCR Internalization Assay
The PathHunter™ eXpress CXCR2 Activated GPCR Internalization Assay measures CXCR2 (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.
Marca: DiscoverX 93-0934E3CP5M

Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The PathHunter™ CXCR2 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. CXCL8/IL-8) based internalization of CXCR2 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged CXCR2. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting CXCR2-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.Especificaciones
β-Arrestin-2 recruitment | |
Recombinant Human IL-8 (CXCL8) | |
CXCR2 | |
CXCR2 |
Assay-Ready eXpress Kits | |
200 dp (2 x 96-well) | |
U2OS |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
DiscoverX PathHunter™ eXpress CXCR2 Activated GPCR Internalization Assay
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido