Learn More
DiscoverX PathHunter™ eXpress P2RY2 Activated GPCR Internalization Assay
Click to view available options
Cantidad:
200 dp (2 x 96-pocillos)
Tamaño de la unidad:
1 set
Descripción
The PathHunter™ P2RY2 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. UTP, ATP) based internalization of P2RY2 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged P2RY2. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting P2RY2-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | GPCR |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | P2RY2 |
| Objetivo | P2RY2 |