missing translation for 'onlineSavingsMsg'
Learn More

Pax5/BSAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

353.00€ - 552.00€

Especificaciones

Antígeno Pax5/BSAP
Dilución Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
18652466
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-38790-25ul
25 μL
353.00€
25 microlitros
View alternative products
missing translation for 'toSaveUpto' 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
18009843
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-38790
0.1 mL
552.00€
0.10 ml
View alternative products
missing translation for 'toSaveUpto' 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

Pax5/BSAP Polyclonal specifically detects Pax5/BSAP in Human, Rat, Hamster samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Pax5/BSAP
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Rat, Hamster
Q02548
5079
This antibody was developed against a recombinant protein corresponding to amino acids: SGILGITSPSADTNKRKRDEGIQESPVPNGHSLPGRDFLRKQMRGDLFTQQQL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
Cancer, Transcription Factors and Regulators
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5
PAX5
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.