missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX5 Monoclonal Antibody (SP34), Invitrogen™
Rabbit Monoclonal Antibody
Marca: Invitrogen MA516389
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.Especificaciones
PAX5 | |
Monoclonal | |
0.067 | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
Q02548 | |
PAX5 | |
Synthetic peptide from C-terminus of human Pax-5 protein | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
SP34 | |
Unconjugated | |
PAX5 | |
ALL3; B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; EBB-1; KLP; paired box 5; paired box gene 5; paired box gene 5 (B-cell lineage specific activator); paired box homeotic gene 5; paired box protein Pax-5; paired domain gene 5; Pax5; Pax-5; Pax5 variant 1; Pax5 variant 2; RGD1565236; transcription factor PAX 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
500453, 5079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |