Learn More
PDAP1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP3-20223JF646
Descripción
PDAP1 Polyclonal antibody specifically detects PDAP1 in Human samples. It is validated for Western Blot, Immunofluorescence
Especificaciones
PDAP1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
HASPP28, PAP1PDGFA-associated protein 1, PAP28 kDa heat- and acid-stable phosphoprotein, PDGFA associated protein 1, PDGF-associated protein | |
Produced in rabbits immunized with E. coli-derived Human PDAP1 fragment | |
0.1 mL | |
Signal Transduction | |
11333 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.