missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human PDGFRA (PDGFR alpha), GST Tag Recombinant Protein
Human PDGFRA (PDGFR alpha), GST Tag Recombinant Protein
Marca: Thermo Scientific™ PV3811
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family.Especificaciones
NP_006197 | |
5156 | |
10 μg | |
PDGFR Kinase Family | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
PDGFRA | |
Baculovirus | |
Recombinant, Catalytic Domain, Cytoplasmic | |
Receptor Tyrosine Kinases |
Kinase Assay | |
89.8 kDa | |
PDGFRA (PDGFR α) | |
aa 550-1089 | |
Recombinant | |
Storage Buffer: 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
platelet-derived growth factor receptor, alpha polypeptide | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |