Learn More
PDGFRB & PTPN11 Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Marca:  Abnova DI0034.1Set
Código de producto. 16131074
  • 677.00€ / 1 set
Descripción
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the PDGFRB protein, and the other against the PTPN11 protein for use in in situ Proximity Ligation Assay.
Especificaciones
Especificaciones
PDGFRB
Unconjugated
CD140B/JTK12/PDGF-R-beta/PDGFR/PDGFR1
1 Set
Primary
Antibody pair set content: 1. PDGFRB rabbit purified polyclonal antibody (20 ug) 2. PTPN11 mouse purified polyclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
In situ PLA
platelet-derived growth factor receptor, beta polypeptide
PDGFRB
RUO
5159/5781
Human
Ab Pair