Learn More
PDGFRB & PTPN11 Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Marca: Abnova DI0034.1Set
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the PDGFRB protein, and the other against the PTPN11 protein for use in in situ Proximity Ligation Assay.Especificaciones
| PDGFRB | |
| Unconjugated | |
| CD140B/JTK12/PDGF-R-beta/PDGFR/PDGFR1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. PDGFRB rabbit purified polyclonal antibody (20 ug) 2. PTPN11 mouse purified polyclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| platelet-derived growth factor receptor, beta polypeptide | |
| PDGFRB | |
| RUO | |
| 5159/5781 | |
| Human | |
| Ab Pair |