missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDPK1 Monoclonal Antibody (3H3D9), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA531835
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-31835 has been tested in indirect ELISA.
PDPK1 (3 Phosphoinositide Dependent Protein Kinase 1) phosphorylates AGC kinases. PDPK1 activates conventional PKC and PKC zeta through phosphorylation of critical threonine residues in the activation loop. PDPK1 also phosphorylates Protein Kinase B (PKB) at threonine 308 in the presence of phosphatidylinositol-3,4,5-trisphosphate. Active Akt inactivates Glycogen Synthase Kinase 3 (GSK3), eventually leading to the dephosphorylation and activation of glycogen synthase, and the stimulation of glycogen synthesis. Because of the role that PDPK1 plays in insulin-induced glycogen synthesis and PKC activation, it is a potentially important target for metabolic drug research.Especificaciones
PDPK1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O15530 | |
Pdpk1 | |
Purified recombinant fragment of human PDPK1 (AA: 457-556 expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
3H3D9 | |
Unconjugated | |
Pdpk1 | |
3-phosphoinositide dependent protein kinase 1; 3-phosphoinositide dependent protein kinase-1; 3-phosphoinositide-dependent protein kinase 1; 3-phosphoinositide-dependent protein kinase 2 pseudogene; HGNC:8809; hPDK1; isoenzyme 1; kinase isoenzyme 1; lipoamide; mPDK1; OTTHUMP00000174525; OTTHUMP00000205076; PDK1; PDPK1; PDPK2; PDPK2P; Pkb kinase; PkB kinase like gene 1; PkB-like 1; PRO0461; Protein kinase B kinase | |
Mouse | |
Protein G | |
RUO | |
5170 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |