missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PDZK2 Recombinant Protein
Marca: Abnova™ H00079849-P01.25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH29042 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
80.1 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MEKAADLQDTASLTLKFKFNPKLGIDNPVLSLAEDHDPYDPWSLERPRFCLLSKEEGKSFGFHLQQELGRAGHVVCRVDPGTSAQRQGLQEGDRILAVNNDVVEHEDYAVVVRRIRASSPRVLLTVLARHAHDVARAQLGEDAHLCPTLGPGVRPRLCHIVKDEGGFGFSVTHGNQGPFWLVLSTGGAAERAGVPPGARLLEVNGLWQSGQQVTLLVAGPEVEEQCRQLGLPLAAPLAEGWALPTKPRCLHLEKGPQGFGFLLREEKGLDGRPGQFLWEVDPGLPAKKAGMQAGDRLVAVAGESVEGLGHEETVSRIQGQGSCVSLTVVDPEADRFFSMVRLSPLLFLENTEAPASPQGSSSASLVETEDPSLEDTSVPSVPLGSRQCFLYPGPGGSYGFRLSCVASGPRLFISQVTPGGSAARAGLQVGDVILEVNGYPVGGQNDLERLQQLPEAEPPLCLKLAARSLRGLEAWIPPGAAEDWALASDLL | |
FLJ22756/IKEPP/PDZK2 | |
PDZD3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
79849 | |
PDZK2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PDZD3 | |
Human | |
Recombinant | |
Solution |