missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEDF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA129882
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: RIPA lysates from Y79 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PEDF is a noninhibitory serpin with neurotrophic, anti-angiogenic, and anti-tumorigenic properties. It is a 50 kDa glycoprotein produced and secreted in many tissues throughout the body. A major component of the anti-angiogenic action of PEDF is the induction of apoptosis in proliferating endothelial cells. In addition, PEDF is able to inhibit the activity of angiogenic factors, such as VEGF and FGF-2. The neuroprotective effects of PEDF are achieved through suppression of neuronal apoptosis induced by peroxide, glutamate, or other neurotoxins. The recent identification of a lipase-linked cell membrane receptor for PEDF (PEDF-R) that binds to PEDF with high affinity (1) should facilitate further elucidation of the underlying mechanisms of this pluripotent serpin. To date, PEDF-R is the only signaling receptor known to be used by a serpin family member. The unique range of PEDF activities implicate it as a potential therapeutic agent for the treatment of vasculature-related neurodegenerative diseases, such as age-related macular degeneration (AMD) and proliferative diabetic retinopathy (PDR). PEDF also has the potential to be useful in the treatment of various angiogenesis-related diseases including a number of cancers.Especificaciones
PEDF | |
Polyclonal | |
Unconjugated | |
SERPINF1 | |
AI195227; alpha-2 antiplasmin; caspin; Cell proliferation-inducing gene 35 protein; Dmrs91; EPC 1; EPC1; EPC-1; OI12; OI6; PEDF; Pedfl; PIG 35; PIG35; pigment epithelium derived factor; pigment epithelium-derived factor; Sdf3; SDF-3; serine (or cysteine) peptidase inhibitor, clade F, member 1; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1; serine (or cysteine) proteinase inhibitor, clade F), member 1; serine (or cysteine) proteinase inhibitor, clade F, member 1; serpin F1; serpin family F member 1; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1; serpin peptidase inhibitor, clade F, member 1; SERPINF 1; Serpinf1; serpin-F1; sserpin family F member 1; stromal cell derived factor 3; stromal cell-derived factor 3; testis tissue sperm-binding protein Li 70n | |
Rabbit | |
Protein A | |
RUO | |
20317, 287526, 5176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 0.15M NaCl, 30% glycerol and 0.05% sodium azide; pH 7.4 | |
P36955, P97298 | |
SERPINF1 | |
His tagged fusion protein corresponding to full length human PEDF (Pigment Epithelial Derived Factor) expressed in baby hamster kidney cells. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |