Learn More
3M™ Auriculares PELTOR™ WS™ Alert™ XP con Bluetooth™
Los auriculares 3M™ PELTOR™ WS™ Alert™ XP con Bluetooth™ permiten que los usuarios mantengan conversaciones mediante telefonía inalámbrica o escuchen música, incluso en entornos ruidosos.
Marca: 3M™ 7100020054
Detalles adicionales : Peso : 5.00000kg
Descripción
- Stay protected whilst taking calls wirelessly or listening to the radio - even in noisy environments
- Level dependent function lets the user hear surrounding noise and converse with colleagues
- Fast, easy turning of radio stations
- Bluetooth™ for wireless hands-free calls or music streaming
- Advanced speaker system that provides full-bodied dynamic sound
- Battery charge via cable plugged into electrical socket or USB
- Fast Tuning
- Ghost voice guided interface
- Memory/preset function
Especificaciones
Bluetooth™ Headset | |
Built-in FM Radio Receiver, Adjustable Damping Function | |
29 dB |