Learn More
Gilac™ Perforated Crate
Perforated Crate is a stacking perforated crate ideal for storing delicate foodstuffs such as fruit and vegetables; it provides ventilation and is highly resistant to intensive use.
Marca: Gilac™ G048244
Detalles adicionales : Peso : 1.54000kg
Descripción
- Stacking perforated crate.
- Ideal for storing all products requiring air circulation.
- Highly resistant for intensive use.
- Ideal for storing fruit and vegetables.
- Easy to grab hold of thanks to practical ergonomic integrated handles.
- Solid container lids are also suitable for perforated containers.
- Compatible with all GILAC 4 and 6-wheel dollies.
Especificaciones
8.5 gal. | |
None | |
HDPE | |
Crate | |
500 x 300 x 300 mm | |
19.7 in. | |
11.8 in. | |
11.8 in. |
32 L | |
Red | |
Rectangular | |
Yes | |
500 mm | |
300 mm | |
300 mm |