Learn More
Gilac™ Perforated Crate
Perforated Crate is a stacking perforated crate ideal for storing delicate foodstuffs such as fruit and vegetables; it provides ventilation and is highly resistant to intensive use.
Marca: Gilac™ G179577
Detalles adicionales : Peso : 1.49000kg
Descripción
- Stacking perforated crate.
- Ideal for storing all products requiring air circulation.
- Highly resistant for intensive use.
- Ideal for storing fruit and vegetables.
- Easy to grab hold of thanks to practical ergonomic integrated handles.
- Solid container lids are also suitable for perforated containers.
- Compatible with all GILAC 4 and 6-wheel dollies.
Especificaciones
6.6 gal. | |
None | |
HDPE | |
Crate | |
600 x 400 x 135 mm | |
23.6 in. | |
15.7 in. | |
5.3 in. |
25 L | |
Gray | |
Rectangular | |
Yes | |
600 mm | |
400 mm | |
135 mm |