Learn More
Crison™ Electrodos de pH
Marca: Crison™ 50151
Detalles adicionales : Peso : 0.10000kg
Especificaciones
Measures samples with proteins and sudden changes in temperature or alkaline; ideal for research | |
Glass | |
-10°C to +100°C |
Diaphragm:Ceramic, 3; Electrolyte: CRISOLYT G | |
0 to 14 |