missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Caveolin 2 (Ser23) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA564546
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Phospho-Caveolin 2 (Ser23) Polyclonal Antibody detects endogenous levels of Caveolin 2 only when phosphorylated at Ser23.
Caveolae are specialized domains of the plasma membrane that are implicated in the sequestration of a variety of lipid and protein molecules. It has been suggested that these important cellular organelles have a pivotal role in such diverse biochemical processes as lipid metabolism, growth regulation, signal transduction, and apoptosis. Caveolin interacts with and regulates heterotrimeric G-proteins. Currently, there are three members of the caveolin multigene family which are known to encode 21-24 kDa integral membrane proteins that comprise the major structural component of the caveolar membrane in vivo. Caveolin-2 protein is abundantly expressed in fibroblasts and differentiated adipocytes, smooth and skeletal muscle, and endothelial cells. The expression of caveolin-1 is similar to that of caveolin-2 while caveolin-3 expression appears to be limited to muscle tissue types.
Especificaciones
Phospho-Caveolin 2 (Ser23) | |
Polyclonal | |
Unconjugated | |
CAV2 | |
AI447843; CAV; CAV2; caveolae; caveolae protein, 20-kD; caveolin 2; caveolin 2 isoform a and b; caveolin-2; MGC12294; Phospho-CAV2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12390, 363425, 858 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P51636, Q2IBC5, Q9WVC3 | |
CAV2 | |
A synthesized peptide derived from human Caveolin 2 (Phospho-Ser23) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Phospho-Caveolin 2 (Ser23) Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido