missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-FOXO4 (Ser197) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA537648
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
A suggested positive control for Western blot is 293 cells; suggested positive control for IHC is human breast carcinoma; suggested positive control for ICC/IF is MCF-7 cells.
Phospho-FOXO4 (Ser197) Polyclonal antibody specifically detects Phospho-FOXO4 (Ser197) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
Phospho-FOXO4 (Ser197) | |
Polyclonal | |
Unconjugated | |
FOXO4 | |
AFX; Afx1; Afxh; AFXzeta; Fkhr3; Fork head domain transcription factor AFX1; forkhead box O4; forkhead box protein O4; forkhead protein; FOXO4; FOXO4b; MGC120490; MLLT7; myeloid/lymphoid or mixed lineage-leukemia translocation to 7 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7; OTTHUMP00000023497; RGD1561201 | |
Rabbit | |
Affinity chromatography | |
RUO | |
302415, 4303, 54601 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P98177, Q9WVH3 | |
FOXO4 | |
Peptide sequence around phosphorylation site of serine 197 (A-A-S(p)-M-D) derived from Human AFX. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |