missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PKC alpha (Thr497) Recombinant Rabbit Monoclonal Antibody (PKCaT497-F1), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537116
429.35 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
The PKC family of serine/threonine kinases, including PRKCA (PKC alpha), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II, and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. Protein kinase C (PKC) can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This kinase has been reported to play roles in many different cellular processes, such as cell adhesion, cell transformation, cell cycle checkpoint, and cell volume control. Knockout studies in mice suggest that this kinase may be a fundamental regulator of cardiac contractility and Ca(2+) handling in myocytes.
Especificaciones
Phospho-PKC alpha (Thr497) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide | |
P05696, P17252, P20444 | |
PRKCA | |
A synthetic phospho-peptide corresponding to residues surrounding Thr497 of human phospho PKC alpha. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG κ |
Flow Cytometry, Western Blot | |
PKCaT497-F1 | |
Unconjugated | |
PRKCA | |
AAG6; aging-associated gene 6; AI875142; EC 2.7.11.13; FHC; FTH; FTHL6; im:7139045; kinase PKC-alpha; KPCA; MGC104426; PIG15; PKC III; Pkca; PKC-A; PKC-alpha; PKC-III; PKRCA; PLIF; PRKACA; Prkca; prkcaa; protein kinase C; protein kinase C alpha; protein kinase C alpha type; protein kinase C alpha type; protein kinase C, alpha, a; protein kinase C, alpha; protein kinase C, alpha, a; protein kinase c-alpha; si:dkey-8l19.1; Unknown (protein for IMAGE:7139045); unnamed protein product | |
Rabbit | |
Protein A/G | |
RUO | |
18750, 24680, 5578 | |
-20°C | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Phospho-PKC alpha (Thr497) Recombinant Rabbit Monoclonal Antibody (PKCaT497-F1), Invitrogen™ > 200 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido